6-Bromo-5-fluoro-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

129341-5G 129341-1G 129341-25G 129341-500MG
77633-990EA 298 USD
77633-990 77636-514 77653-424 77648-526
6-Bromo-5-fluoro-1-methyl-1H-indazole ≥97%
6-Bromo-5-fluoro-1-methyl-1H-indazole

Precaución: For Research Use Only

Formula: C₈H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD18089817
CAS Number: 1286734-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR