6-Fluoro-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

129329-5G 129329-1G 129329-25G 129329-500MG
77631-558EA 1187 USD
77631-558 77636-512 77657-354 77648-524
6-Fluoro-2-methyl-2H-indazole ≥97%
6-Fluoro-2-methyl-2H-indazole

Precaución: For Research Use Only

Formula: 150.153 g/mol
MW: C₈H₇FN₂
Storage Temperature: Ambient
MDL Number: MFCD11977538
CAS Number: 348-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR