4-Bromo-7-fluoro-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

129315-5G 129315-1G 129315-500MG
77653-422EA 2485 USD
77653-422 77640-540 77644-512
4-Bromo-7-fluoro-2-methyl-2H-indazole ≥97%
4-Bromo-7-fluoro-2-methyl-2H-indazole

Precaución: For Research Use Only

Formula: 229.049 g/mol
MW: C₈H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD32670518
CAS Number: 2411637-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR