4-Bromo-7-fluoro-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

129314-5G 129314-25G 129314-1G 129314-500MG
77631-556EA 1315 USD
77631-556 77657-352 77640-538 77644-510
4-Bromo-7-fluoro-1-methyl-1H-indazole ≥97%
4-Bromo-7-fluoro-1-methyl-1H-indazole

Precaución: For Research Use Only

Formula: 229.049 g/mol
MW: C₈H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD30183582
CAS Number: 2090912-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR