1,2-Dimethoxy-4-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

129255-5G 129255-25G 129255-1G 129255-500MG
77633-988EA 494 USD
77633-988 77653-420 77640-534 77648-520
1,2-Dimethoxy-4-ethylbenzene ≥97%
1,2-Dimethoxy-4-ethylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00673001
CAS Number: 5888-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR