2-Fluoro-1-iodo-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

129231-5G 129231-1G 129231-500MG
77653-418EA 1868 USD
77653-418 77640-530 77648-516
2-Fluoro-1-iodo-3-methoxybenzene ≥95%
2-Fluoro-1-iodo-3-methoxybenzene

Precaución: For Research Use Only

Formula: 252.025 g/mol
MW: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD18396915
CAS Number: 1080673-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR