1-(2,5-Difluorophenyl)cycloheptanol ≥97%

Supplier: AOBChem USA

Warning

128283-5G 128283-25G 128283-1G 128283-500MG
77631-194EA 1599.59 USD
77631-194 77657-342 77644-508 77648-510
1-(2,5-Difluorophenyl)cycloheptanol ≥97%
1-(2,5-Difluorophenyl)cycloheptanol

Precaución: For Research Use Only

Formula: C₁₃H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD17223835
CAS Number: 1341277-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR