3-Hydroxy-2,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

128145-1G 128145-5G 128145-500MG
77636-506EA 695 USD
77636-506 77657-338 77648-508
3-Hydroxy-2,6-dimethoxybenzaldehyde ≥97%
3-Hydroxy-2,6-dimethoxybenzaldehyde

Precaución: For Research Use Only

Formula: 182.173 g/mol
MW: C₉H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD34827521
CAS Number: 80832-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR