3-Cyano-2-methoxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

127983-5G 127983-1G 127983-500MG
77657-336EA 2142 USD
77657-336 77640-526 77648-506
3-Cyano-2-methoxyisonicotinic acid ≥95%
3-Cyano-2-methoxyisonicotinic acid

Precaución: For Research Use Only

Formula: 178.145 g/mol
MW: C₈H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28779598
CAS Number: 1807036-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR