2-Iodo-6-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

127692-5G 127692-500MG 127692-25G 127692-1G
77631-190EA 1484.23 USD
77631-190 77652-320 77657-334 77640-524
2-Iodo-6-isopropoxypyridine ≥95%
2-Iodo-6-isopropoxypyridine

Precaución: For Research Use Only

Formula: C₈H₁₀INO
Storage Temperature: Ambient
MDL Number: MFCD34827230
CAS Number: 2921872-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR