2-(Difluoromethoxy)-4,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

127428-5G 127428-1G 127428-500MG
77631-188EA 1484.23 USD
77631-188 77636-500 77644-502
2-(Difluoromethoxy)-4,5-dimethylbenzaldehyde ≥95%
2-(Difluoromethoxy)-4,5-dimethylbenzaldehyde

Precaución: For Research Use Only

Formula: 200.182 g/mol
MW: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20502019
CAS Number: 1354960-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR