3-(tert-Butoxymethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

12686-5G 12686-25G
77633-986EA 62 USD
77633-986 77653-410
3-(tert-Butoxymethyl)phenylboronic acid ≥95%
3-(tert-Butoxymethyl)phenylboronic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD03788411
CAS Number: 858364-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR