4'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

126826-5G 126826-1G 126826-25G
77633-984EA 228 USD
77633-984 77636-496 77657-330
4'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥97%
4'-(Dimethylamino)-2,2,2-trifluoroacetophenone

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00099468
CAS Number: 2396-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR