(3-Fluoro-2-methylphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

126336-5G 126336-25G 126336-1G 126336-500MG
77631-550EA 1381 USD
77631-550 77657-326 77640-514 77648-498
(3-Fluoro-2-methylphenyl)(phenyl)methanone ≥95%
(3-Fluoro-2-methylphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₄H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD34661843
CAS Number: 142153-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR