1-(3-Fluoro-2-methylphenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

126330-5G 126330-1G 126330-25G 126330-500MG
77631-180EA 1484.23 USD
77631-180 77636-494 77653-404 77644-496
1-(3-Fluoro-2-methylphenyl)cyclopentanol ≥97%
1-(3-Fluoro-2-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 194.245 g/mol
MW: C₁₂H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD34826493
CAS Number: 2921841-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR