1-(3-Fluoro-2-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

126326-5G 126326-1G 126326-25G 126326-500MG
77631-548EA 1231 USD
77631-548 77636-492 77653-402 77644-494
1-(3-Fluoro-2-methylphenyl)propan-1-ol ≥95%
1-(3-Fluoro-2-methylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 168.208 g/mol
MW: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD11521246
CAS Number: 1270584-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR