1-(2,4-Difluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

126183-5G 126183-25G 126183-1G 126183-500MG
77631-546EA 1218 USD
77631-546 77653-400 77640-512 77644-492
1-(2,4-Difluorophenyl)propan-1-ol ≥95%
1-(2,4-Difluorophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 172.172 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD12787299
CAS Number: 1247998-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR