3'-(Trifluoromethyl)propiophenone ≥95%
Supplier: AOBChem USA
|
Warning
|
12587-5G
12587-1G
12587-25G
12587-500MG
77631-176EA
1484.23
USD
77631-176
77636-488
77653-396
77648-496
3'-(Trifluoromethyl)propiophenone ≥95%
3'-(Trifluoromethyl)propiophenone
Precaución: For Research Use Only
Formula:
C₁₀H₉F₃O Storage Temperature: Ambient |
MDL Number:
MFCD00039227 CAS Number: 1533-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...