3-(Methylsulfanyl)benzenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Methylsulphanyl)phenol

125727-5G 125727-25G 125727-1G
77633-978EA 194 USD
77633-978 77653-394 77640-510
3-(Methylsulfanyl)benzenol ≥97%
3-(Methylsulfanyl)benzenol

Precaución: For Research Use Only

Formula: C₇H₈OS
MW: 140.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11207488
CAS Number: 3463-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR