Benzo[b]thiophen-2-yl(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

125609-5G 125609-25G 125609-1G 125609-500MG
77631-174EA 1519.25 USD
77631-174 77635-428 77640-508 77648-492
Benzo[b]thiophen-2-yl(phenyl)methanol ≥95%
Benzo[b]thiophen-2-yl(phenyl)methanol

Precaución: For Research Use Only

Formula: 240.32 g/mol
MW: C₁₅H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD00971587
CAS Number: 116496-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR