2,2-Difluoro-1-(thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

125568-1G 125568-5G 125568-500MG
77636-484EA 560 USD
77636-484 77657-320 77648-490
2,2-Difluoro-1-(thiophen-2-yl)ethanone ≥97%
2,2-Difluoro-1-(thiophen-2-yl)ethanone

Precaución: For Research Use Only

Formula: 162.157 g/mol
MW: C₆H₄F₂OS
Storage Temperature: Ambient
MDL Number: MFCD16618121
CAS Number: 2991-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR