2-Bromo-3-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

125519-5G 125519-25G
77633-974EA 71 USD
77633-974 77657-318
2-Bromo-3-iodobenzoic acid ≥97%
2-Bromo-3-iodobenzoic acid

Precaución: For Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036150
CAS Number: 855198-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR