2,4-Dibromo-5-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

125418-5G 125418-25G 125418-1G 125418-500MG
77631-170EA 1484.23 USD
77631-170 77653-390 77640-506 77648-486
2,4-Dibromo-5-fluoro-N-methoxy-N-methylbenzamide ≥97%
2,4-Dibromo-5-fluoro-N-methoxy-N-methylbenzamide

Precaución: For Research Use Only

Formula: C₉H₈Br₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD34826163
CAS Number: 2918964-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR