2-Bromo-6-(2,2,2-trifluoroethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

125272-5G 125272-1G 125272-500MG
77631-166EA 1484.23 USD
77631-166 77640-502 77648-482
2-Bromo-6-(2,2,2-trifluoroethoxy)pyridine ≥95%
2-Bromo-6-(2,2,2-trifluoroethoxy)pyridine

Precaución: For Research Use Only

Formula: 256.02 g/mol
MW: C₇H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD13189518
CAS Number: 869640-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR