(2-Bromo-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

125249-5G 125249-1G 125249-500MG
77653-386EA 2143 USD
77653-386 77644-482 77644-484
(2-Bromo-6-methoxyphenyl)(methyl)sulfane ≥97%
(2-Bromo-6-methoxyphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₈H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD35166121
CAS Number: 2411991-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR