1-(2,5-Dibromopyridin-4-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

124616-5G 124616-1G 124616-500MG 124616-25G
77631-160EA 1484.23 USD
77631-160 77636-474 77652-316 77657-314
1-(2,5-Dibromopyridin-4-yl)cyclopentanol ≥95%
1-(2,5-Dibromopyridin-4-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 321.008 g/mol
MW: C₁₀H₁₁Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD34825910
CAS Number: 2918955-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR