1-(5-Bromo-2-chloropyridin-4-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

124577-5G 124577-25G 124577-1G 124577-500MG
77631-156EA 1484.23 USD
77631-156 77657-312 77640-496 77644-480
1-(5-Bromo-2-chloropyridin-4-yl)cyclopentanol ≥97%
1-(5-Bromo-2-chloropyridin-4-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 276.557 g/mol
MW: C₁₀H₁₁BrClNO
Storage Temperature: Ambient
MDL Number: MFCD34825896
CAS Number: 2918950-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR