1-(5-Chloro-2,3-difluoropyridin-4-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

124571-5G 124571-1G 124571-25G 124571-500MG
77631-154EA 1565.6 USD
77631-154 77636-472 77657-310 77644-478
1-(5-Chloro-2,3-difluoropyridin-4-yl)cyclopentanol ≥97%
1-(5-Chloro-2,3-difluoropyridin-4-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 233.642 g/mol
MW: C₁₀H₁₀ClF₂NO
Storage Temperature: Ambient
MDL Number: MFCD34825894
CAS Number: 2969309-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR