1-(6-Bromo-2-fluoropyridin-3-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

124508-5G 124508-25G 124508-1G 124508-500MG
77631-152EA 1707.74 USD
77631-152 77653-380 77640-494 77644-476
1-(6-Bromo-2-fluoropyridin-3-yl)cyclopentanol ≥95%
1-(6-Bromo-2-fluoropyridin-3-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 260.103 g/mol
MW: C₁₀H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34825883
CAS Number: 2918969-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR