6-Bromo-3-cyclopentyl-2-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

124472-1G 124472-5G 124472-500MG
77633-972EA 964 USD
77633-972 77653-378 77648-476
6-Bromo-3-cyclopentyl-2-fluorobenzaldehyde ≥95%
6-Bromo-3-cyclopentyl-2-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: 271.125 g/mol
MW: C₁₂H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD28388022
CAS Number: 2169355-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR