3-Amino-5-iodo-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

124060-500MG 124060-5G 124060-1G
77652-314EA 557 USD
77652-314 77657-306 77640-492
3-Amino-5-iodo-4-methylbenzoic acid ≥95%
3-Amino-5-iodo-4-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₈H₈INO₂
Storage Temperature: Ambient
CAS Number: 215229-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR