3-Amino-5-chloro-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

124053-1G 124053-5G 124053-500MG
77636-470EA 510 USD
77636-470 77653-376 77648-474
3-Amino-5-chloro-4-methylbenzoic acid ≥95%
3-Amino-5-chloro-4-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD11557528
CAS Number: 1092307-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR