3-Chloro-2-fluoro-4-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

12355-5G 12355-25G 12355-1G 12355-500MG
77631-148EA 1484.23 USD
77631-148 77653-374 77640-490 77644-472
3-Chloro-2-fluoro-4-(trimethylsilyl)pyridine ≥95%
3-Chloro-2-fluoro-4-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: C₈H₁₁ClFNSi
MW: 203.717 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21099528
CAS Number: 851386-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR