1-(6-Bromo-2,3-difluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

123512-5G 123512-25G 123512-1G 123512-500MG
77631-146EA 1777.78 USD
77631-146 77653-372 77640-488 77644-470
1-(6-Bromo-2,3-difluorophenyl)propan-1-ol ≥95%
1-(6-Bromo-2,3-difluorophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 251.068 g/mol
MW: C₉H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34825416
CAS Number: 2969308-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR