1-(3-Bromo-2,6-difluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

123502-5G 123502-1G 123502-500MG
77631-144EA 1777.78 USD
77631-144 77640-486 77644-468
1-(3-Bromo-2,6-difluorophenyl)propan-1-ol ≥95%
1-(3-Bromo-2,6-difluorophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₉H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34825410
CAS Number: 1692508-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR