1-Cyclobutyl-2,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

123499-5G 123499-1G 123499-25G 123499-500MG
77631-142EA 1823.1 USD
77631-142 77636-466 77653-370 77648-472
1-Cyclobutyl-2,4-difluorobenzene ≥97%
1-Cyclobutyl-2,4-difluorobenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂
Storage Temperature: Ambient
MDL Number: MFCD34825407
CAS Number: 2918961-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR