1-(4-Fluoro-2-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

123472-5G 123472-25G 123472-1G 123472-500MG
77631-540EA 1152 USD
77631-540 77657-302 77640-484 77648-470
1-(4-Fluoro-2-methoxyphenyl)ethanol ≥95%
1-(4-Fluoro-2-methoxyphenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11520990
CAS Number: 1339915-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR