4-[2-(2-Bromophenoxy)ethyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

123336-5G 123336-1G 123336-500MG 123336-25G
77633-968EA 363 USD
77633-968 77636-464 77652-312 77653-368
4-[2-(2-Bromophenoxy)ethyl]morpholine ≥97%
4-[2-(2-Bromophenoxy)ethyl]morpholine

Precaución: For Research Use Only

Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD01308474
CAS Number: 101558-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR