1-Bromo-3-chloro-2,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

123197-5G 123197-25G
77633-966EA 108 USD
77633-966 77657-300
1-Bromo-3-chloro-2,5-difluorobenzene ≥97%
1-Bromo-3-chloro-2,5-difluorobenzene

Precaución: For Research Use Only

Formula: C₆H₂BrClF₂
MW: 227.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845991
CAS Number: 1160573-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR