4-(Cyclopropylmethoxy)-2,6-difluorobenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
122998-5G
122998-25G
122998-1G
122998-500MG
77631-136EA
1777.78
USD
77631-136
77635-424
77636-460
77644-462
4-(Cyclopropylmethoxy)-2,6-difluorobenzoic acid ≥95%
4-(Cyclopropylmethoxy)-2,6-difluorobenzoic acid
Precaución: For Research Use Only
Formula:
228.192 g/mol MW: C₁₁H₁₀F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD34825166 CAS Number: 2918959-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...