4-Bromo-1-tosyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

122611-5G 122611-25G 122611-1G
77633-962EA 214 USD
77633-962 77657-296 77640-476
4-Bromo-1-tosyl-1H-pyrazole ≥97%
4-Bromo-1-tosyl-1H-pyrazole

Precaución: For Research Use Only

Formula: C₁₀H₉BrN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD06080375
CAS Number: 116228-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR