2-Bromo-3-fluoro-4-methoxybenzonitrile ≥95%

Supplier: AOBChem USA

Warning

122599-1G 122599-5G 122599-500MG
77633-960EA 844 USD
77633-960 77657-294 77648-464
2-Bromo-3-fluoro-4-methoxybenzonitrile ≥95%
2-Bromo-3-fluoro-4-methoxybenzonitrile

Precaución: For Research Use Only

Formula: 230.034 g/mol
MW: C₈H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD26687963
CAS Number: 1501465-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR