1-(Methoxymethoxy)-2,4-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

12198-5G 12198-25G 12198-1G 12198-500MG
77633-958EA 556 USD
77633-958 77653-360 77640-474 77648-462
1-(Methoxymethoxy)-2,4-bis(trifluoromethyl)benzene ≥95%
1-(Methoxymethoxy)-2,4-bis(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 274.16 g/mol
MW: C₁₀H₈F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD32631763
CAS Number: 2364584-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR