1-Iodo-4-(4-(trifluoromethoxy)phenoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

121890-5G 121890-25G 121890-1G 121890-500MG
77631-538EA 688 USD
77631-538 77653-358 77640-472 77648-460
1-Iodo-4-(4-(trifluoromethoxy)phenoxy)benzene ≥95%
1-Iodo-4-(4-(trifluoromethoxy)phenoxy)benzene

Precaución: For Research Use Only

Formula: 380.101 g/mol
MW: C₁₃H₈F₃IO₂
Storage Temperature: Ambient
MDL Number: MFCD14582284
CAS Number: 873203-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR