(4-(4-(Trifluoromethoxy)phenoxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

121886-5G 121886-25G 121886-1G 121886-500MG
77631-536EA 1088 USD
77631-536 77653-356 77640-470 77644-458
(4-(4-(Trifluoromethoxy)phenoxy)phenyl)methanol ≥95%
(4-(4-(Trifluoromethoxy)phenoxy)phenyl)methanol

Precaución: For Research Use Only

Formula: 284.231 g/mol
MW: C₁₄H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD29058619
CAS Number: 1686102-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR