4-(4-(Trifluoromethoxy)phenoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

121885-5G 121885-1G 121885-25G 121885-500MG
77631-534EA 1227 USD
77631-534 77636-458 77653-354 77648-458
4-(4-(Trifluoromethoxy)phenoxy)benzaldehyde ≥95%
4-(4-(Trifluoromethoxy)phenoxy)benzaldehyde

Precaución: For Research Use Only

Formula: 282.215 g/mol
MW: C₁₄H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28991462
CAS Number: 137736-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR