5-Bromo-2,2-difluoro-6-iodobenzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

121837-5G 121837-25G 121837-1G 121837-500MG
77631-130EA 1484.23 USD
77631-130 77635-420 77636-456 77644-456
5-Bromo-2,2-difluoro-6-iodobenzo[d][1,3]dioxole ≥97%
5-Bromo-2,2-difluoro-6-iodobenzo[d][1,3]dioxole

Precaución: For Research Use Only

Formula: 362.895 g/mol
MW: C₇H₂BrF₂IO₂
Storage Temperature: Ambient
CAS Number: 1770849-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR