(4-Bromo-3-chlorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

121659-5G 121659-1G 121659-25G 121659-500MG
77631-530EA 1381 USD
77631-530 77636-452 77653-352 77648-454
(4-Bromo-3-chlorophenyl)(thiomorpholino)methanone ≥97%
(4-Bromo-3-chlorophenyl)(thiomorpholino)methanone

Precaución: For Research Use Only

Formula: 320.633 g/mol
MW: C₁₁H₁₁BrClNOS
Storage Temperature: Ambient
MDL Number: MFCD23030116
CAS Number: 1500741-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR