(4-Bromo-3-chlorophenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
121656-5G
121656-1G
121656-25G
121656-500MG
77631-528EA
1441
USD
77631-528
77636-450
77653-348
77644-454
(4-Bromo-3-chlorophenyl)(pyrrolidin-1-yl)methanone ≥97%
(4-Bromo-3-chlorophenyl)(pyrrolidin-1-yl)methanone
Precaución: For Research Use Only
Formula:
288.568 g/mol MW: C₁₁H₁₁BrClNO Storage Temperature: Ambient |
MDL Number:
MFCD23027207 CAS Number: 1506099-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...