4-Bromo-3-chloro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

121652-5G 121652-1G 121652-25G 121652-500MG
77631-526EA 1189 USD
77631-526 77636-448 77657-290 77644-452
4-Bromo-3-chloro-N-methylbenzamide ≥97%
4-Bromo-3-chloro-N-methylbenzamide

Precaución: For Research Use Only

Formula: C₈H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD24018816
CAS Number: 1517784-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR